ENA282548581 Enamine

heptane-1-sulfonyl chloride

  • MDL number MFCD12783398
  • Cas Number : 927-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-61276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61276
Availability details for
Quantity
  • Availability
  • Add to Cart