ENA282548582 Enamine

3-(4-chlorophenyl)-6-hydroxy-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD01665939
  • Cas Number : 28921-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61277

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61277
Availability details for
Quantity
  • Availability
  • Add to Cart