ENA282548583 Enamine

2-[2-chloro-4-(chlorosulfonyl)phenoxy]acetic acid

  • MDL number MFCD09938465
  • Cas Number : 1017055-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61278
Availability details for
Quantity
  • Availability
  • Add to Cart