ENA282548586 Enamine

5-cyclopropyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD10006870
  • Cas Number : 1017477-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61282
Availability details for
Quantity
  • Availability
  • Add to Cart