ENA282548588 Enamine

3-(3,6-dioxo-1,2,3,6-tetrahydropyridazin-1-yl)benzoic acid

  • MDL number MFCD00567620
  • Cas Number : 87565-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 312...314
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60355

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60355
Availability details for
Quantity
  • Availability
  • Add to Cart