ENA282548590 Enamine

4,6-dimethyl-N-[(3-methyl-1H-pyrazol-4-yl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxamide

  • MDL number MFCD12903882
  • Cas Number : 1182778-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61287
Availability details for
Quantity
  • Availability
  • Add to Cart