ENA282548599 Enamine

4-phenoxypyridin-3-amine

  • MDL number MFCD11184593
  • Cas Number : 132038-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61298
Availability details for
Quantity
  • Availability
  • Add to Cart