ENA282548600 Enamine

1-(2,6-dichlorophenyl)-5-ethyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD12106596
  • Cas Number : 1154206-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61300
Availability details for
Quantity
  • Availability
  • Add to Cart