ENA282548602 Enamine

4-[2-(2-methoxyethoxy)acetamido]cyclohexane-1-carboxylic acid

  • MDL number MFCD12607497
  • Cas Number : 1183863-89-9
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61302
Availability details for
Quantity
  • Availability
  • Add to Cart