ENA282548606 Enamine

1-(2-methoxyethyl)-1H-indol-5-amine

  • MDL number MFCD11106362
  • Cas Number : 1152866-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61306
Availability details for
Quantity
  • Availability
  • Add to Cart