ENA282548615 Enamine

1-(4-aminophenyl)piperidine-4-carboxamide

  • MDL number MFCD09737672
  • Cas Number : 223786-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61334
Availability details for
Quantity
  • Availability
  • Add to Cart