ENA282548618 Enamine

[1-(4-aminobenzoyl)piperidin-4-yl]methanol

  • MDL number MFCD11132986
  • Cas Number : 1094233-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61338
Availability details for
Quantity
  • Availability
  • Add to Cart