ENA282548620 Enamine

5-bromo-7-chloro-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD11207508
  • Cas Number : 1094288-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61340
Availability details for
Quantity
  • Availability
  • Add to Cart