ENA282548621 Enamine

methyl 3-(4-aminophenoxy)propanoate

  • MDL number MFCD12779273
  • Cas Number : 130198-72-0
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60360
Availability details for
Quantity
  • Availability
  • Add to Cart