ENA282548622 Enamine

5-amino-2-(thiomorpholin-4-yl)benzoic acid

  • MDL number MFCD12816215
  • Cas Number : 78243-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263...265
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61344

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61344
Availability details for
Quantity
  • Availability
  • Add to Cart