ENA282548627 Enamine

2-({[5-(thiophen-2-yl)-1,2-oxazol-3-yl]methyl}sulfanyl)acetic acid

  • MDL number MFCD08445240
  • Cas Number : 923721-92-0
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60363
Availability details for
Quantity
  • Availability
  • Add to Cart