ENA282548631 Enamine

3-amino-N-(pyrazin-2-yl)benzamide

  • MDL number MFCD12090941
  • Cas Number : 1156412-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61358
Availability details for
Quantity
  • Availability
  • Add to Cart