ENA282548632 Enamine

4-bromo-2-methoxybenzamide

  • MDL number MFCD12913570
  • Cas Number : 812667-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-139
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61359
Availability details for
Quantity
  • Availability
  • Add to Cart