ENA282548634 Enamine

2,2,2-trifluoroethyl N-(3-fluorophenyl)carbamate

  • MDL number MFCD00091248
  • Cas Number : 200876-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61364
Availability details for
Quantity
  • Availability
  • Add to Cart