ENA282548635 Enamine

2,2,2-trifluoroethyl N-[2-(trifluoromethyl)phenyl]carbamate

  • MDL number MFCD11099769
  • Cas Number : 1087788-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61365

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61365
Availability details for
Quantity
  • Availability
  • Add to Cart