ENA282548636 Enamine

2,2,2-trifluoroethyl N-(2,5-dichlorophenyl)carbamate

  • MDL number MFCD11099872
  • Cas Number : 1087798-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61366
Availability details for
Quantity
  • Availability
  • Add to Cart