ENA282548639 Enamine

5-bromo-2-fluorobenzene-1-sulfonyl chloride

  • MDL number MFCD09763690
  • Cas Number : 339370-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61369
Availability details for
Quantity
  • Availability
  • Add to Cart