ENA282548640 Enamine

2-(3-bromophenyl)morpholine

  • MDL number MFCD10035261
  • Cas Number : 1018612-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61370
Availability details for
Quantity
  • Availability
  • Add to Cart