ENA282548641 Enamine

1-amino-3-(cyclopropylmethoxy)propan-2-ol

  • MDL number MFCD11136682
  • Cas Number : 1019546-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61371
Availability details for
Quantity
  • Availability
  • Add to Cart