ENA282548643 Enamine

N-(1-cyclopropylethyl)-4-methoxyaniline

  • MDL number MFCD11147338
  • Cas Number : 1020962-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-61377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61377
Availability details for
Quantity
  • Availability
  • Add to Cart