ENA282548644 Enamine

1-[2-(4-ethylpiperazin-1-yl)-5-fluorophenyl]ethan-1-amine

  • MDL number MFCD11137149
  • Cas Number : 1019553-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61378
Availability details for
Quantity
  • Availability
  • Add to Cart