ENA282548647 Enamine

4-methyl-3-(trifluoromethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD15209709
  • Cas Number : 151258-36-5
  • Molecular Weight : 258.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61383

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-61383
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart