ENA282548650 Enamine

2-[hydroxy(4-nitrophenyl)methyl]prop-2-enenitrile

  • MDL number MFCD09030957
  • Cas Number : 127489-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61386
Availability details for
Quantity
  • Availability
  • Add to Cart