ENA282548655 Enamine

5-methyl-2-(1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD11127760
  • Cas Number : 133662-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61399
Availability details for
Quantity
  • Availability
  • Add to Cart