ENA282548660 Enamine

1-(2-chlorophenyl)-5-cyclopropyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD12106504
  • Cas Number : 1154372-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61405
Availability details for
Quantity
  • Availability
  • Add to Cart