ENA282548662 Enamine

5,8-dichloro-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD00956375
  • Cas Number : 22406-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61408
Availability details for
Quantity
  • Availability
  • Add to Cart