ENA282548666 Enamine

4-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}aniline

  • MDL number MFCD09048226
  • Cas Number : 926216-36-6
  • Molecular Weight : 230.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61414
Availability details for
Quantity
  • Availability
  • Add to Cart