ENA282548670 Enamine

1-(pyridin-2-ylmethyl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD12804851
  • Cas Number : 1184416-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61419

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61419
Availability details for
Quantity
  • Availability
  • Add to Cart