ENA282548673 Enamine

5-[(3-bromophenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD08444641
  • Cas Number : 923737-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54542

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54542
Availability details for
Quantity
  • Availability
  • Add to Cart