ENA282548686 Enamine

4-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzene-1-carboximidamide dihydrochloride

  • MDL number MFCD14705832
  • Cas Number : 1221722-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60393
Availability details for
Quantity
  • Availability
  • Add to Cart