ENA282548687 Enamine

[(2-fluorophenyl)sulfanyl]formonitrile

  • MDL number MFCD15209720
  • Cas Number : 2605-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61501
Availability details for
Quantity
  • Availability
  • Add to Cart