ENA282548688 Enamine

2-[2-(propan-2-yloxy)phenyl]acetic acid

  • MDL number MFCD09925031
  • Cas Number : 171814-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61521
Availability details for
Quantity
  • Availability
  • Add to Cart