ENA282548694 Enamine

quinoline-8-carbohydrazide

  • MDL number MFCD01365835
  • Cas Number : 85949-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60433

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60433
Availability details for
Quantity
  • Availability
  • Add to Cart