ENA282548701 Enamine

2-chloro-N-[4-(3-chlorophenoxy)phenyl]acetamide

  • MDL number MFCD11961068
  • Cas Number : 69838-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-60438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60438
Availability details for
Quantity
  • Availability
  • Add to Cart