ENA282548703 Enamine

2-amino-N-(2-methylpropyl)acetamide hydrochloride

  • MDL number MFCD13562468
  • Cas Number : 175154-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60440
Availability details for
Quantity
  • Availability
  • Add to Cart