ENA282548707 Enamine

ethyl 3-amino-5-(3-bromophenyl)thiophene-2-carboxylate

  • MDL number MFCD09835656
  • Cas Number : 1094398-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55066

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55066
Availability details for
Quantity
  • Availability
  • Add to Cart