ENA282548708 Enamine

methyl 3-(1H-indol-3-yl)-2-(naphthalen-2-ylformamido)propanoate

  • MDL number MFCD01257137
  • Cas Number : 1396968-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60445
Availability details for
Quantity
  • Availability
  • Add to Cart