ENA282548714 Enamine

1-(4-{[1-(2,3-dihydro-1-benzofuran-5-yl)ethyl]amino}piperidin-1-yl)ethan-1-one

  • MDL number MFCD13801361
  • Cas Number : 1157628-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60452
Availability details for
Quantity
  • Availability
  • Add to Cart