ENA282548717 Enamine

4-hydroxy-6,8-dimethoxyquinoline-3-carboxylic acid

  • MDL number MFCD05228452
  • Cas Number : 874498-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 299...301
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55122

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55122
Availability details for
Quantity
  • Availability
  • Add to Cart