ENA282548725 Enamine

3-methyl-2-(2-oxopyrrolidin-1-yl)butanoic acid

  • MDL number MFCD11214951
  • Cas Number : 1094652-93-3
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60474
Availability details for
Quantity
  • Availability
  • Add to Cart