ENA292595904 Enamine

4-ethylpiperazine-1-carboxamide hydrochloride

  • MDL number MFCD16039981
  • Cas Number : 1235441-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-49765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49765
Availability details for
Quantity
  • Availability
  • Add to Cart