ENA292595906 Enamine

2,2,2-trifluoroethyl N-[6-(2-methoxyethoxy)pyridin-3-yl]carbamate

  • MDL number MFCD16039984
  • Cas Number : 1235438-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55209
Availability details for
Quantity
  • Availability
  • Add to Cart