ENA292595907 Enamine

2,2,2-trifluoroethyl N-(2-chloro-4-nitrophenyl)carbamate

  • MDL number MFCD16039985
  • Cas Number : 1235439-39-0
  • Molecular Weight : 298.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55212
Availability details for
Quantity
  • Availability
  • Add to Cart