ENA292595910 Enamine

2,2,2-trifluoroethyl N-[3-(2-oxo-1,3-oxazolidin-3-yl)phenyl]carbamate

  • MDL number MFCD16039989
  • Cas Number : 1235438-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56803
Availability details for
Quantity
  • Availability
  • Add to Cart