ENA292595913 Enamine

N-[4-(aminomethyl)phenyl]-2,2,2-trifluoroacetamide

  • MDL number MFCD16039992
  • Cas Number : 1235441-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57038
Availability details for
Quantity
  • Availability
  • Add to Cart